Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAG-3 Antibody (C9B7W), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB212271 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB212271 0.125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB212271 Supplier Novus Biologicals Supplier No. NB10063601AF594
Add to Cart
Add to Cart

Rat Monoclonal Antibody

LAG-3 Monoclonal antibody specifically detects LAG-3 in Mouse samples. It is validated for Flow Cytometry

Specifications

Antigen LAG-3
Applications Flow Cytometry
Classification Monoclonal
Clone C9B7W
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias CD223, CD223 antigen, LAG3, LAG-3, lymphocyte activating 3, lymphocyte activation gene 3 protein, lymphocyte-activation gene 3, Secreted lymphocyte activation gene 3 protein, sLAG-3
Host Species Rat
Immunogen This LAG-3 Antibody (C9B7W) was prepared from murine CD223 Ig fusion protein
Purification Method Protein G purified
Quantity 0.125 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cancer, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3902
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.