Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAG-3 (CD223) Mouse anti-Human, APC-R700, Clone: T47-530, BD

Catalog No. BDB565774 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565774 100 Tests
BDB565775 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB565774 Supplier BD Biosciences Supplier No. 565774
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The T47-530 specifically recognizes the Lymphocyte Activation Gene 3 (LAG-3) protein which is also known as, Protein FDC, or CD223. LAG-3 is a ñ70 kDa type I transmembrane glycoprotein that belongs to the Ig superfamily and exhibits homology to CD4. LAG-3 is expressed on NK cells, regulatory T cells, and activated conventional T cells with higher expression found on CD8+ T cells compared with CD4+ T cells. LAG-3 is an activation induced cell surface molecule that like CD4, binds MHC class II molecules, but with much higher affinity. This may enable LAG-3 to act as a negative competitor of CD4 for MHC class II ligand binding. LAG-3 may associate with the TCR-CD3 complex to downregulate TCR signal transduction and T cell clonal expansion. In contrast, LAG-3-induced signaling may promote dendritic cell activation.

This antibody was conjugated to BD Horizon APC-R700, which has been developed exclusively by BD Biosciences as a better alternative to Alexa Fluor™ 700. APC-R700 excites and emits at similar wavelengths to Alexa Fluor™ 700 yet exhibits significantly improved brightness. This dye can be excited by the red laser and detected with the same filter set as Alexa Fluor™ (eg, 730/45nm filter). APC-R700 is a tandem fluorochrome that combines APC with R700, a proprietary organic dye. This dye has been developed exclusively by BD Biosciences as a brighter alternative to Alexa Fluor™ 700. Due to similar excitation and emission properties, APC-R700 and Alexa Fluor™ 700 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LAG-3 (CD223)
Classification Monoclonal
Clone T47-530
Conjugate APC-R700
Formulation Aqueous buffered solution containing BSA, protein stabilizer, glycerol and ≤0.09% sodium azide.
Gene Alias LAG3, CD223, FDC, Lymphocyte activation gene 3 protein, Protein FDC
Host Species Mouse
Immunogen Human LAG-3 Recombinant Protein
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.