Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LAG-3 (CD223) Mouse anti-Human, PE, Clone: T47-530, BD Pharmingen™
Mouse Monoclonal Antibody
$173.40 - $398.50
Specifications
| Antigen | LAG-3 (CD223) |
|---|---|
| Clone | T47-530 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | PE |
Description
LAG-3 (CD223) Monoclonal antibody specifically detects LAG-3 (CD223) in Human samples. It is validated for Flow cytometry.
LAG-3 (CD223) Monoclonal antibody specifically detects LAG-3 (CD223) in Human samples. It is validated for Flow cytometry.Specifications
| LAG-3 (CD223) | |
| Flow Cytometry | |
| PE | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human LAG-3 Recombinant Protein | |
| Primary |
| T47-530 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| LAG3; CD223; FDC; Lymphocyte activation gene 3 protein; Protein FDC | |
| IgG1 κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions and unconjugated antibody and free PE were removed. |
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title