Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAG-3 Mouse anti-Human, Clone: 17B4, Novus Biologicals™
SDP

Catalog No. N197657100U Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N197657100U 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N197657100U Supplier Novus Biologicals Supplier No. NBP197657100UG

Mouse Monoclonal Antibody

LAG-3 Monoclonal specifically detects LAG-3 in Human, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Block/Neutralize.

Specifications

Antigen LAG-3
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone 17B4
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 5 μ/mL, Flow Cytometry, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 10 μ/mL, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:150
Formulation Liquid. In PBS containing 10% glycerol with 0.02% sodium azide
Gene Alias 17b4, 17b4 lag3, 17b4 neutralizing, anti human lag3, Anti-LAG-3, CD223, CD223 Antigen, FDC, Human LAG3, Human LAG-3, LAG-3, LAG-3 17b4, LAG-3 blocking, lymphocyte activation gene 3 protein, lymphocyte-activation gene 3, Protein FDC
Gene Symbols LAG3
Host Species Mouse
Immunogen Synthetic peptide corresponding to 30 aa (GPPAAAPGHPLAPGPHPAAPSSWGPRPRRY) from the first N-terminal D1 domain of human LAG-3 (lymphocyte activation gene-3).
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 3902
Test Specificity Recognizes the 30 aa extra-loop of the first N-terminal D1 domain of human LAG-3.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.