Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAIR1 Antibody (113), PE, Novus Biologicals™
SDP

Catalog No. NBP143324PE Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP143324PE 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP143324PE Supplier Novus Biologicals Supplier No. NBP143324PE
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

Ensure accurate, reproducible results in Western Blot, Flow Cytometry, Immunoprecipitation

LAIR1 Monoclonal specifically detects LAIR1 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen LAIR1
Applications Flow Cytometry
Classification Monoclonal
Clone 113
Conjugate PE
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias CD305 antigen, CD305leukocyte-associated Ig-like receptor 1, hLAIR1, LAIR-1, leukocyte-associated immunoglobulin-like receptor 1
Gene Symbols LAIR1
Host Species Armenian Hamster
Immunogen The immunogen for this antibody was CD305.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3903
Target Species Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.