Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAIR1 Antibody (342219), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB364588 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB364588 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB364588 Supplier Novus Biologicals Supplier No. FAB2664W
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LAIR1 Monoclonal antibody specifically detects LAIR1 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF

Specifications

Antigen LAIR1
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 342219
Conjugate DyLight 650
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD305 antigen, CD305leukocyte-associated Ig-like receptor 1, hLAIR1, LAIR-1, leukocyte-associated immunoglobulin-like receptor 1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human LAIR1 isoform 1, Gln22-His163, Accession # Q6GTX8
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3903
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.