Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAIR1 Antibody (NKTA255), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB002592 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002592 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002592 Supplier Novus Biologicals Supplier No. NBP262214G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LAIR1 Monoclonal specifically detects LAIR1 in Human samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen LAIR1
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone NKTA255
Conjugate DyLight 488
Dilution Western Blot, Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CD305 antigen, CD305leukocyte-associated Ig-like receptor 1, hLAIR1, LAIR-1, leukocyte-associated immunoglobulin-like receptor 1
Gene Symbols LAIR1
Host Species Mouse
Immunogen Activated NK cells and CD3- thymocytes
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3903
Test Specificity The mouse monoclonal antibody NKTA255 recognizes CD305/ LAIR1, a 40 kDa type I transmembrane glycoprotein expressed on NK, T, and B cells, monocytes, dendritic cells, eosinophils, basophils, mast cells, CD34+ hematopoietic progenitor cells and thymocytes.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.