Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAIR2 Antibody (319701), FITC, Novus Biologicals™
SDP

Catalog No. NB304325 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB304325 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB304325 Supplier Novus Biologicals Supplier No. FAB2665F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LAIR2 Monoclonal antibody specifically detects LAIR2 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF

Specifications

Antigen LAIR2
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 319701
Conjugate FITC
Dilution Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias CD306 antigen, CD306leukocyte-associated Ig-like receptor-2, LAIR-2, leukocyte-associated Ig-like receptor 2, leukocyte-associated immunoglobulin-like receptor 2, MGC71634
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human LAIR2 isoform 1, Gln22-Pro152, Accession # Q6ISS4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3904
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.