Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Lalistat 2

Click to view available options
Quantity:
10 mg
50 mg
Description
Lalistat 2 is a selective lysosomal acid lipase inhibitor (IC50 = 152 nM). Exhibits no inhibition of human pancreatic lipase or bovine milk lipoprotein lipase (up to 10 μM).
Specifications
Specifications
Chemical Name or Material | 4-(Piperidin-1-yl)-1,2,5-thiadiazol-3-yl piperidine-1-carboxylate |
CAS | 1234569-09-5 |
Assay Percent Range | >98% |
Quantity | 10 mg |
Molecular Formula | C13H20N4O2S |
Formula Weight | Observed MW: 296.39 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction