Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lambda Light Chain Antibody (1-155-2), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP179124IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP179124IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP179124IR Supplier Novus Biologicals Supplier No. NBP179124IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lambda Light Chain Monoclonal specifically detects Lambda Light Chain in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Lambda Light Chain
Applications Flow Cytometry
Classification Monoclonal
Clone 1-155-2
Conjugate DyLight 755
Dilution Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias IGLC, immunoglobulin lambda constant group
Gene Symbols IGLC1
Host Species Mouse
Immunogen Human IgA1 lambda myeloma protein
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3536
Test Specificity The mouse monoclonal antibody 1-155-2 recognizes lambda light chains (22.5 kDa) of human immunoglobulin.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.