Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lambda Light Chain Antibody (1-155-2), PE/Atto594, Novus Biologicals™
SDP

Catalog No. N179124P594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N179124P594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N179124P594 Supplier Novus Biologicals Supplier No. NBP179124PEATT594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lambda Light Chain Monoclonal specifically detects Lambda Light Chain in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Lambda Light Chain
Applications Flow Cytometry
Classification Monoclonal
Clone 1-155-2
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias IGLC, immunoglobulin lambda constant group
Gene Symbols IGLC1
Host Species Mouse
Immunogen Human IgA1 lambda myeloma protein
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3537.0
Test Specificity The mouse monoclonal antibody 1-155-2 recognizes lambda light chains (22.5 kDa) of human immunoglobulin.
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.