Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lambda Light Chain Antibody (LcN-2), Novus Biologicals™
SDP

Catalog No. NBP2627520 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
200 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2627520 200 μg
NBP26275120 20 μg
NBP26275110 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP2627520 Supplier Novus Biologicals Supplier No. NBP262751200UG

Mouse Monoclonal Antibody

LambdaLightChain Monoclonal antibody specifically detects LambdaLightChain in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen LambdaLightChain
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone LcN-2
Conjugate Unconjugated
Dilution Western Blot 0.5-1ug/ml, Flow Cytometry 0.5ug-1ug/million cells in 0.1ml, Immunocytochemistry/Immunofluorescence 1-2ug/ml, Immunohistochemistry-Paraffin 0.25-0.5ug/ml
Formulation 10mM PBS with 0.05% BSA with 0.05% Sodium Azide
Gene Alias IGLC, immunoglobulin lambda constant group
Gene Symbols IGL
Host Species Mouse
Immunogen Purified human IgG
Purification Method Protein A or G purified
Quantity 200 μg
Primary or Secondary Primary
Gene ID (Entrez) 3536
Target Species Human, Mammalia
Content And Storage Store at 4°C.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.