Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lambda Light Chain Antibody (SPM559), PerCP, Novus Biologicals™
SDP

Catalog No. NBP234424P Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP234424P 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP234424P Supplier Novus Biologicals Supplier No. NBP234424PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lambda Light Chain Monoclonal specifically detects Lambda Light Chain in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Lambda Light Chain
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SPM559
Conjugate PerCP
Dilution Flow Cytometry
Gene Alias IGLC, immunoglobulin lambda constant group
Gene Symbols IGLC1
Host Species Mouse
Immunogen Purified human IgG
Molecular Weight of Antigen 22.5 kDa
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3536
Test Specificity This monoclonal antibody is specific to lambda light chain of immunoglobulin and shows no cross-reaction with lambda light chain or any of the five heavy chains. In mammals, the two light chains in an antibody are always identical, with only one type of light chain, kappa or lambda. The ratio of Kappa to Lambda is 70:30. However, with the occurrence of multiple myeloma or other B-cell malignancies this ratio is disturbed. Antibody to the lambda light chain is reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkins lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is malignant.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.