Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lambda5/IGLL1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | Lambda5/IGLL1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
Lambda5/IGLL1 Polyclonal specifically detects Lambda5/IGLL1 in Human samples. It is validated for Western Blot.Specifications
Lambda5/IGLL1 | |
Polyclonal | |
Purified | |
RUO | |
P15814 | |
3543 | |
Synthetic peptides corresponding to IGLL1(immunoglobulin lambda-like polypeptide 1) The peptide sequence was selected from the N terminal of IGLL1. Peptide sequence RSRWGRFLLQRGSWTGPRCWPRGFQSKHNSVTHVFGSGTQLTVLSQPKAT. | |
Primary |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
14.1, AGM2, CD179 antigen-like family member B, CD179b, CD179b antigen, Ig lambda-5, IGL1, IGL5CD179B, IGLJ14.1, IGLL, IGO, IGVPB, immunoglobulin lambda-like polypeptide 1, Immunoglobulin omega polypeptide, immunoglobulin omega polypeptide chain, immunoglobulin-related 14.1 protein, Immunoglobulin-related protein 14.1, lambda5, Pre-B lymphocyte-specific protein-2, VPREB2 | |
IGLL1 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title