Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lamin A + C Antibody (mab636), Novus Biologicals™
SDP

Catalog No. NB10074451 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10074451 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10074451 Supplier Novus Biologicals Supplier No. NB10074451
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 2 publications

Lamin A + C Monoclonal specifically detects Lamin A + C in Bovine, Human, Mouse, Pig, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot.

Specifications

Antigen Lamin A + C
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone mab636
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Frozen 1:100
Formulation Tissue culture supernatant with 0.05% Sodium Azide
Gene Accession No. Q3ZD69
Gene Alias CDCD1, CDDC, CMD1A, CMT2B1, dilated 1A (autosomal dominant), EMD2, FPLD, HGPSFPL, lamin A/C, lamin A/C-like 1, lamin-A/C, LDP1, LFP, LGMD1B, limb girdle muscular dystrophy 1B (autosomal dominant), LMN1IDC, LMNC, LMNL1, prelamin-A/C, PRO1,70 kDa lamin, progeria 1 (Hutchinson-Gilford type), renal carcinoma antigen NY-REN-32
Gene Symbols LMNA
Host Species Mouse
Immunogen Porcine lamin preparation.
Purification Method Unpurified
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4000
Test Specificity Detects lamin A/C from human, bovine and porcine samples.
Target Species Bovine, Human, Mouse, Pig, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Supernatant
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.