Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lamin A + C R453W Antibody (12A-2F5), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP177401A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP177401A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP177401A94 Supplier Novus Biologicals Supplier No. NBP177401AF594

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Lamin A + C R453W Monoclonal specifically detects Lamin A + C R453W in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Lamin A + C R453W
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 12A-2F5
Conjugate Alexa Fluor 594
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias 70 kDa Lamin, CDCD1, CDDC, Charcot Marie Tooth Disease Axonal Type 2B1, CMD1A, CMT2B1, EMD2, FPL, FPLD, HGPS, IDC, LAMIN A, Lamin A/C, Lamin A/C R453W, LAMIN C, LDP1, LFP, LGMD1B, LMN 1, LMN A, LMN C, LMN1, LMNA, LMNC, PRELAMIN A, PRO1, Progeria 1 (Hutchinson Gilford Type)
Gene Symbols LMNA
Host Species Mouse
Immunogen Synthetic peptide corresponding to amino acids 449-457 of human Lamin A + C [AccessionNo.: NP_733821.1]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 4000
Test Specificity Disease-associated point-mutant R453W of Lamin A + C
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.