Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lamin A + C R453W Antibody (12A-2F5), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB338060 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB338060 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB338060 Supplier Novus Biologicals Supplier No. NBP177401MFV450

Mouse Monoclonal Antibody

Lamin A + C R453W Monoclonal antibody specifically detects Lamin A + C R453W in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation

Specifications

Antigen Lamin A + C R453W
Applications Western Blot, ELISA, Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone 12A-2F5
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias 70 kDa Lamin, CDCD1, CDDC, Charcot Marie Tooth Disease Axonal Type 2B1, CMD1A, CMT2B1, EMD2, FPL, FPLD, HGPS, IDC, LAMIN A, Lamin A/C, Lamin A/C R453W, LAMIN C, LDP1, LFP, LGMD1B, LMN 1, LMN A, LMN C, LMN1, LMNA, LMNC, PRELAMIN A, PRO1, Progeria 1 (Hutchinson Gilford Type)
Host Species Mouse
Immunogen Synthetic peptide corresponding to amino acids 449-457 of human Lamin A + C [Accession#: NP_733821.1]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4000
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.