Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lamin A + C R453W Antibody (12A-2F5), Novus Biologicals™
SDP

Catalog No. NBP177401 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP177401 0.1 mL
NBP177401SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP177401 Supplier Novus Biologicals Supplier No. NBP177401

Mouse Monoclonal Antibody has been used in 1 publication

Lamin A + C R453W Monoclonal specifically detects Lamin A + C R453W in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen Lamin A + C R453W
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone 12A-2F5
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:10-1:500
Gene Accession No. NP_733821.1
Gene Alias 70 kDa Lamin, CDCD1, CDDC, Charcot Marie Tooth Disease Axonal Type 2B1, CMD1A, CMT2B1, EMD2, FPL, FPLD, HGPS, IDC, LAMIN A, Lamin A/C, Lamin A/C R453W, LAMIN C, LDP1, LFP, LGMD1B, LMN 1, LMN A, LMN C, LMN1, LMNA, LMNC, PRELAMIN A, PRO1, Progeria 1 (Hutchinson Gilford Type)
Gene Symbols LMNA
Host Species Mouse
Immunogen Synthetic peptide corresponding to amino acids 449-457 of human Lamin A + C [Accession#: NP_733821.1]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Chromatin Modifiers, Chromatin Research, DNA replication Transcription Translation and Splicing, Nuclear Envelope Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4000
Test Specificity Disease-associated point-mutant R453W of Lamin A + C
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.