Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lamin B Receptor Antibody (BB2SS3F3), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP259947R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2-59947R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP2-59947R Supplier Novus Biologicals Supplier No. NBP259947R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lamin B Receptor Monoclonal specifically detects Lamin B Receptor in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Lamin B Receptor
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone BB2SS3F3
Conjugate DyLight 550
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias DHCR14B, FLJ43126, Integral nuclear envelope inner membrane protein, lamin B receptor, lamin-B receptor, LMN2R, MGC9041, PHA
Gene Symbols LBR
Host Species Mouse
Immunogen GST fused to N-terminal fragment of human Lamin B Receptor (amino acids residues 1-211)
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Cellular Signaling
Primary or Secondary Primary
Gene ID (Entrez) 3930
Test Specificity Specificity of human, mouse Lamin B Receptor antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.