Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lamin B Receptor Antibody (BB2SS3F3), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB308715 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB308715 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB308715 Supplier Novus Biologicals Supplier No. NBP259947MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lamin B Receptor Monoclonal antibody specifically detects Lamin B Receptor in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)

Specifications

Antigen Lamin B Receptor
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone BB2SS3F3
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate
Gene Alias DHCR14B, FLJ43126, Integral nuclear envelope inner membrane protein, lamin B receptor, lamin-B receptor, LMN2R, MGC9041, PHA
Host Species Mouse
Immunogen GST fused to N-terminal fragment of human Lamin B Receptor (amino acids residues 1-211)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Signaling
Primary or Secondary Primary
Gene ID (Entrez) 3930
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.