Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lamin B1 Antibody (CL3929), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 5 publications
Supplier: Novus Biologicals NBP259783
Description
Lamin B1 Monoclonal specifically detects Lamin B1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Lamin B1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
LMNB1 | |
This antibody was developed against a recombinant protein corresponding to amino acids: RTTRGKRKRVDVEESEASSSVSISHSASA | |
100 μL | |
Apoptosis, Cancer, Core ESC Like Genes, Loading Controls, Stem Cell Markers, Tumor Suppressors | |
4001 | |
Human | |
Purified |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
4001 | |
Western Blot 1 μg/mL, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
ADLD, lamin B1, lamin-B1, LMN, LMN2, LMNB, MGC111419 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction