Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lamin B1 Antibody (CL3929), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 5 publications
$428.50 - $694.00
Specifications
Antigen | Lamin B1 |
---|---|
Clone | 4001 |
Dilution | Western Blot 1 μg/mL, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
Lamin B1 Monoclonal specifically detects Lamin B1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Lamin B1 | |
Western Blot 1 μg/mL, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
4001 | |
This antibody was developed against a recombinant protein corresponding to amino acids: RTTRGKRKRVDVEESEASSSVSISHSASA | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
4001 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Apoptosis, Cancer, Core ESC Like Genes, Loading Controls, Stem Cell Markers, Tumor Suppressors | |
ADLD, lamin B1, lamin-B1, LMN, LMN2, LMNB, MGC111419 | |
LMNB1 | |
IgG1 | |
Protein A purified |
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title