Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lamin B2 Antibody (OTI2C6), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP272303A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP272303A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP272303A70 Supplier Novus Biologicals Supplier No. NBP272303AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lamin B2 Monoclonal specifically detects Lamin B2 in Human samples. It is validated for Western Blot.

Specifications

Antigen Lamin B2
Applications Western Blot
Classification Monoclonal
Clone OTI2C6
Conjugate Alexa Fluor 750
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias lamin B2, lamin-B2, LMN2LAMB2, MGC2721
Gene Symbols LMNB2
Host Species Mouse
Immunogen Full length recombinant protein of human Lamin B2 (NP_116126) produced in HEK293T cells.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 84823
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.