Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lamin B2 Antibody (OTI2C6), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB288256 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288256 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288256 Supplier Novus Biologicals Supplier No. NBP272303APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lamin B2 Monoclonal antibody specifically detects Lamin B2 in Human samples. It is validated for Western Blot

Specifications

Antigen Lamin B2
Applications Western Blot
Classification Monoclonal
Clone OTI2C6
Conjugate APC
Dilution Western Blot
Formulation PBS
Gene Alias lamin B2, lamin-B2, LMN2LAMB2, MGC2721
Host Species Mouse
Immunogen Full length human recombinant protein of human LMNB2 (NP_116126) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology, Cytoskeleton Markers
Primary or Secondary Primary
Gene ID (Entrez) 84823
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.