Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lamin B2 Rabbit anti-Human, Mouse, Rat, Clone: S02-9I7, Novus Biologicals™
SDP

Catalog No. NB188460 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB188460 100 μg
NB188461 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB188460 Supplier Novus Biologicals Supplier No. NBP315054100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Lamin B2 Monoclonal antibody specifically detects Lamin B2 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen Lamin B2
Applications Western Blot
Classification Monoclonal
Clone S02-9I7
Conjugate Unconjugated
Formulation 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide
Gene Alias lamin B2, lamin-B2, LMN2LAMB2, MGC2721
Host Species Rabbit
Immunogen A synthetic peptide of human Lamin B2 (Uniprot#: Q03252)
Purification Method Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology, Cytoskeleton Markers
Primary or Secondary Primary
Gene ID (Entrez) 84823
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.