Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaminB2 Mouse anti-Human, Clone: J9-C4, Novus Biologicals™
SDP

Catalog No. NBP266923 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP266923 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP266923 Supplier Novus Biologicals Supplier No. NBP266923
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lamin B2 Monoclonal specifically detects Lamin B2 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen LaminB2
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone J9-C4
Concentration 2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, Immunohistochemistry-Paraffin
Formulation PBS (pH7.4), 0.2% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias lamin B2, lamin-B2, LMN2LAMB2, MGC2721
Gene Symbols LMNB2
Host Species Mouse
Immunogen Synthetic peptide within Human Lamin B2 aa 571-620/ 620. (SwissProt: Q03252 Human)
Purification Method Protein A purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 84823
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.