Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Laminin alpha-1 Monoclonal Antibody (4C7)
GREENER_CHOICE

Catalog No. PIMA121194
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA121194 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA121194 Supplier Invitrogen™ Supplier No. MA121194
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody reacts with the terminal globular domain of the A chain. It does not cross react with laminin from mouse or rat. The staining with this antibody is sensitive to fixation of paraffin tissues and use of proteolytic enzymatic treatment. Recommended positive control: skin tissue.

Laminin antibodies are widely used to label blood vessels and basement membranes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Laminin alpha-1
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 4C7
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Tissue culture supernatant with 0.09% sodium azide
Gene LAMA1
Gene Accession No. P25391
Gene Alias LAMA; LAMA1; Laminin A chain; laminin subunit alpha 1; laminin subunit alpha-1; laminin, alpha 1; Laminin-1 subunit alpha; Laminin-3 subunit alpha; PTBHS; S-LAM alpha; S-LAM-alpha; S-laminin subunit alpha
Gene Symbols LAMA1
Host Species Mouse
Immunogen The A chain of human laminin.
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284217
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.