Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laminin alpha 1 Antibody (775016), PE, Novus Biologicals™
SDP

Catalog No. NB120484 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB120484 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB120484 Supplier Novus Biologicals Supplier No. FAB4187P
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Laminin alpha 1 Monoclonal specifically detects Laminin alpha 1 in Human samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining), Immunocytochemistry.

Specifications

Antigen Laminin alpha 1
Applications CyTOF, Flow Cytometry (Intracellular Staining), Immunocytochemistry
Classification Monoclonal
Clone 775016
Conjugate PE
Dilution Intracellular Staining by Flow Cytometry
Formulation PBS
Gene Alias LAMA, LAMA1 laminin, alpha 1, S-LAM-alpha
Host Species Mouse
Immunogen E. coli-derived recombinant human Laminin alpha 1, Leu22-Met269, Accession # P25391
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284217
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.