Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laminin alpha 2 Antibody (2D4), Novus Biologicals™
SDP

Catalog No. NB387994 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB387994 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB387994 Supplier Novus Biologicals Supplier No. H00003908M0150UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Laminin alpha 2 Monoclonal specifically detects Laminin alpha 2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Sandwich ELISA, Western Blot.

Specifications

Antigen Laminin alpha 2
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Sandwich ELISA, Western Blot
Classification Monoclonal
Clone 2D4
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunohistochemistry 1:10 to 1:500, Immunohistochemistry-Paraffin 1:10 to 1:500, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_000417
Gene Alias Laminin M chain, laminin subunit alpha-2, laminin, alpha 2, Laminin-12 subunit alpha, Laminin-2 subunit alpha, LAMMLaminin-4 subunit alpha, Merosin heavy chain
Host Species Mouse
Immunogen LAMA2 (NP_000417, 3013 a.a. ~ 3122 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. DAGVPGHLCDGQWHKVTANKIKHRIELTVDGNQVEAQSPNPASTSADTNDPVFVGGFPDDLKQFGLTTSIPFRGCIRSLKLTKGTGKPLEVNFAKALELRGVQPVSCPAN
Purification Method IgG purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3908
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.