Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laminin alpha 3/Laminin-5 Antibody (546215), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB349232 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB349232 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB349232 Supplier Novus Biologicals Supplier No. FAB21441MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Laminin alpha 3/Laminin-5 Monoclonal specifically detects Laminin alpha 3/Laminin-5 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot.

Specifications

Antigen Laminin alpha 3/Laminin-5
Applications Flow Cytometry (Intracellular Staining), Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 546215
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Immunohistochemistry, Immunoprecipitation, Intracellular Staining by Flow Cytometry, Immunocytochemistry
Formulation 50mM Sodium Borate
Gene Alias a]3B;BM600;E170;LAMA3;Lama3B;LAMNA;LOCS
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Laminin alpha 3/Laminin-5, aa 21-1713, Accession # NP_000218
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.