Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laminin alpha 4 Antibody (OTI5G8), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP271113A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP271113A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP271113A70 Supplier Novus Biologicals Supplier No. NBP271113AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Laminin alpha 4 Monoclonal specifically detects Laminin alpha 4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Laminin alpha 4
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI5G8
Conjugate Alexa Fluor 750
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias DKFZp686D23145, LAMA3LAMA4*-1, laminin alpha 4 chain, laminin subunit alpha-4, laminin, alpha 4, Laminin-14 subunit alpha, Laminin-8 subunit alpha, Laminin-9 subunit alpha
Gene Symbols LAMA4
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 25-270 of human Laminin alpha 4 (NP_001098676) produced in E.coli.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3910
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.