Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laminin alpha 4 Antibody (OTI5G8) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP271113 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271113 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271113 Supplier Novus Biologicals Supplier No. NBP271113
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Laminin alpha 4 Monoclonal specifically detects Laminin alpha 4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Laminin alpha 4
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI5G8
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:2000, Immunohistochemistry 1:150
Formulation Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
Gene Alias DKFZp686D23145, LAMA3LAMA4*-1, laminin alpha 4 chain, laminin subunit alpha-4, laminin, alpha 4, Laminin-14 subunit alpha, Laminin-8 subunit alpha, Laminin-9 subunit alpha
Gene Symbols LAMA4
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 25-270 of human LAMA4(NP_001098676) produced in E.coli.
Molecular Weight of Antigen 199.8 kDa
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 3910
Reconstitution Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.