Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laminin Antibody (4C7), Novus Biologicals™
SDP

Catalog No. NB12017107 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB12017107 0.5 mL
NB018081 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB12017107 Supplier Novus Biologicals Supplier No. NB12017107
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Laminin Monoclonal specifically detects Laminin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Laminin
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 4C7
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:5-1:20, Immunohistochemistry-Paraffin 1:5-1:20
Formulation PBS (pH 7.4), 0.2% BSA, Tween-20 with 0.05% Sodium Azide
Gene Alias LAMA, Laminin A chain, laminin subunit alpha-1, laminin, alpha 1, Laminin-1 subunit alpha, Laminin-3 subunit alpha, PTBHS, S-LAM alpha, S-laminin subunit alpha
Gene Symbols LAMA1
Host Species Mouse
Immunogen Laminin Antibody (4C7) was made to the A chain of human laminin.
Molecular Weight of Antigen 337 kDa
Purification Method Protein G purified
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Apoptosis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Neuroscience, Signal Transduction, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 284217
Test Specificity Laminin Antibody (4C7) reacts with the terminal globular domain of the A chain. The staining with this is sensitive to fixation of paraffin tissues and use of proteolytic enzymatic treatment.
Target Species Human
Content And Storage Store at 4°C.
Form Supernatant
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.