Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laminin beta 3 Antibody (OTI3A2), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP271115R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271115R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271115R Supplier Novus Biologicals Supplier No. NBP271115R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Laminin beta 3 Monoclonal specifically detects Laminin beta 3 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Laminin beta 3
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI3A2
Conjugate DyLight 550
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias BM600-125KDA, Epiligrin subunit bata, FLJ99565, Kalinin B1 chain, Kalinin subunit beta, kalinin-140kDa, LAM5, Laminin 5, Laminin B1k chain, laminin S B3 chain, laminin subunit beta-3, laminin, beta 3, laminin, beta 3 (nicein (125kD), kalinin (140kD), BM600 (125kD)), Laminin-5 subunit beta, LAMNB1, Nicein subunit beta, nicein-125kDa
Gene Symbols LAMB3
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 424-773 of human Laminin beta 3 (NP_000219) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3914.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.