Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laminin gamma 1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP159144 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP159144 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP159144 Supplier Novus Biologicals Supplier No. NBP159144
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Laminin gamma 1 Polyclonal specifically detects Laminin gamma 1 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen Laminin gamma 1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. P11047
Gene Alias LAMB2Laminin-7 subunit gamma, Laminin B2 chain, laminin subunit gamma-1, laminin, gamma 1 (formerly LAMB2), Laminin-1 subunit gamma, Laminin-10 subunit gamma, Laminin-11 subunit gamma, Laminin-2 subunit gamma, Laminin-3 subunit gamma, Laminin-4 subunit gamma, Laminin-6 subunit gamma, Laminin-8 subunit gamma, Laminin-9 subunit gamma, MGC87297, S-LAM gamma, S-laminin subunit gamma
Gene Symbols LAMC1
Host Species Rabbit
Immunogen Synthetic peptides corresponding to LAMC1(laminin, gamma 1 (formerly LAMB2)) The peptide sequence was selected from the middle region of LAMC1. Peptide sequence KTREAQQALGSAAADATEAKNKAHEAERIASAVQKNATSTKAEAERTFAE.
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cytoskeleton Markers, Extracellular Matrix, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 3915
Test Specificity This product is specific to Subunit or Isoform: gamma-1.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Zebrafish
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.