Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Laminin gamma 1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | Laminin gamma 1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
Laminin gamma 1 Polyclonal specifically detects Laminin gamma 1 in Human, Mouse samples. It is validated for Western Blot.Specifications
Laminin gamma 1 | |
Polyclonal | |
Rabbit | |
Apoptosis, Cancer, Cytoskeleton Markers, Extracellular Matrix, Tumor Suppressors | |
LAMB2Laminin-7 subunit gamma, Laminin B2 chain, laminin subunit gamma-1, laminin, gamma 1 (formerly LAMB2), Laminin-1 subunit gamma, Laminin-10 subunit gamma, Laminin-11 subunit gamma, Laminin-2 subunit gamma, Laminin-3 subunit gamma, Laminin-4 subunit gamma, Laminin-6 subunit gamma, Laminin-8 subunit gamma, Laminin-9 subunit gamma, MGC87297, S-LAM gamma, S-laminin subunit gamma | |
LAMC1 | |
IgG | |
This product is specific to Subunit or Isoform: gamma-1. |
Western Blot | |
Unconjugated | |
RUO | |
P11047 | |
3915 | |
Synthetic peptides corresponding to LAMC1(laminin, gamma 1 (formerly LAMB2)) The peptide sequence was selected from the middle region of LAMC1. Peptide sequence KTREAQQALGSAAADATEAKNKAHEAERIASAVQKNATSTKAEAERTFAE. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title