Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAMP-1/CD107a Antibody (508921), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB303406 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB303406 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB303406 Supplier Novus Biologicals Supplier No. FAB4800V

Mouse Monoclonal Antibody

LAMP-1/CD107a Monoclonal antibody specifically detects LAMP-1/CD107a in Human samples. It is validated for Immunohistochemistry, Flow Cytometry, Immunocytochemistry, CyTOF

Specifications

Antigen LAMP-1/CD107a
Applications Immunohistochemistry, Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 508921
Conjugate Alexa Fluor 405
Dilution Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD107 antigen-like family member A, CD107a, CD107a antigen, LAMP-1, LAMPA, LGP120, lysosomal-associated membrane protein 1, lysosome-associated membrane glycoprotein 1, Lysosome-associated membrane protein 1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human LAMP1/CD107a, Ala28-Asn380, Accession # P11279
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cholesterol Metabolism, Endosome Markers, Glycobiology, Lysosome Markers, Mast Cell Markers, Membrane Trafficking and Chaperones, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 3916
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.