Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAMP-2/CD107b Antibody (GL2A7), Novus Biologicals™
SDP

Rat Monoclonal Antibody

$268.00 - $743.50

Specifications

Antigen LAMP-2/CD107b
Clone GL2A7
Dilution Western Blot, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10-1:500
Applications Immunofluorescence, Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB018791
SDP
View Documents
Novus Biologicals
NBP1974990.025ML
0.025 mL
Each for $268.00
Only null left
Add to Cart
 
NBP19749905
SDP
View Documents
Novus Biologicals
NBP1974990.05ML
0.05 mL
Each for $391.50
Only null left
Add to Cart
 
NBP19749902
SDP
View Documents
Novus Biologicals
NBP1974990.2ML
0.2 mL
Each for $743.50
Only null left
Add to Cart
 
Description

Description

LAMP-2/CD107b Monoclonal specifically detects LAMP-2/CD107b in Mouse, Rabbit samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications

Specifications

LAMP-2/CD107b
Western Blot, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10-1:500
Monoclonal
Ascites
RUO
Ascites with No Preservative
CD107 antigen-like family member B, CD107b, CD107b antigen, LAMP-2, LAMPB, LGP110, lysosomal-associated membrane protein 2, lysosome-associated membrane glycoprotein 2, Lysosome-associated membrane protein 2
LAMP2
IgG
Unpurified
GL2A7
Immunofluorescence, Immunocytochemistry, Immunoprecipitation, Western Blot
Unconjugated
Rat
Mouse, Rabbit
P13473, P17047
3920
Mouse liver lysosomal membrane preparation.
Primary
Store at -20C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.