Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAMP-2/CD107b Antibody (GL2A7), Novus Biologicals™
SDP

Catalog No. NB018791 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.05 mL
0.2 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018791 0.025 mL
NBP19749905 0.05 mL
NBP19749902 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB018791 Supplier Novus Biologicals Supplier No. NBP1974990.025ML
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

LAMP-2/CD107b Monoclonal specifically detects LAMP-2/CD107b in Mouse, Rabbit samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen LAMP-2/CD107b
Applications Immunofluorescence, Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone GL2A7
Conjugate Unconjugated
Dilution Western Blot, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10-1:500
Formulation Ascites with No Preservative
Gene Accession No. P13473, P17047
Gene Alias CD107 antigen-like family member B, CD107b, CD107b antigen, LAMP-2, LAMPB, LGP110, lysosomal-associated membrane protein 2, lysosome-associated membrane glycoprotein 2, Lysosome-associated membrane protein 2
Gene Symbols LAMP2
Host Species Rat
Immunogen Mouse liver lysosomal membrane preparation.
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3920
Target Species Mouse, Rabbit
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.