Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAMP-2/CD107b Antibody (H4B4), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP222217R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222217R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222217R Supplier Novus Biologicals Supplier No. NBP222217R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LAMP-2/CD107b Monoclonal specifically detects LAMP-2/CD107b in Human, Mouse, Chicken, Rat (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockout Validated.

Specifications

Antigen LAMP-2/CD107b
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone H4B4
Conjugate DyLight 550
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Gene Accession No. P13473
Gene Alias CD107 antigen-like family member B, CD107b, CD107b antigen, LAMP-2, LAMPB, LGP110, LGP-96, Lysosomal Associated Membrane Protein 2, lysosomal-associated membrane protein 2, lysosome-associated membrane glycoprotein 2, Lysosome-associated membrane protein
Gene Symbols LAMP2
Host Species Mouse
Immunogen LAMP-2/CD107b Antibody (H4B4) was made using a human adherent spleen cells.
Molecular Weight of Antigen 45 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Cognition and Behavior, Growth and Development, Immunology, Innate Immunity, Lipid and Metabolism, Lysosome Markers, Mast Cell Markers, Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 3920
Test Specificity Human. Does not react with Mouse or Rat.
Target Species Human, Mouse, Rat (Negative)
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.