Learn More
Description
Specifications
Specifications
| Antigen | LAMP-2/CD107b |
| Applications | Flow Cytometry, Immunoassay |
| Classification | Monoclonal |
| Clone | H4B4 |
| Conjugate | PE-Cyanine5.5 |
| Dilution | Flow Cytometry, Knockout Validated |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Accession No. | P13473, P13473 |
| Gene Alias | CD107 antigen-like family member B, CD107b, CD107b antigen, LAMP-2, LAMPB, LGP110, LGP-96, Lysosomal Associated Membrane Protein 2, lysosomal-associated membrane protein 2, lysosome-associated membrane glycoprotein 2, Lysosome-associated membrane protein |
| Gene Symbols | LAMP2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
