Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LAMP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | LAMP3 |
|---|---|
| Concentration | 0.25 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-8129
![]()
|
Proteintech Group Inc
126321AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Lysosome-associated membrane glycoprotein 3 (LAMP3, Lamp3) is a protein that in humans is encoded by theLAMP3 gene. This gene is mapped to chromosome 3q26.3-q27. It is one of the lysosome-associated membrane glycoproteins. Dendritic cells (DCs) are the most potent antigen-presenting cells.Specifications
| LAMP3 | |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q5XI99, Q7TST5, Q9UQV4 | |
| 239739, 27074, 303801 | |
| LAMP3 Fusion Protein Ag3325 | |
| Primary | |
| -20°C | |
| LAMP3 |
| 0.25 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| CD208, DC LAMP, DCLAMP, LAMP, LAMP 3, LAMP3, Protein TSC403, TSC403 | |
| LAMP3 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title