Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAMTOR1 Antibody, Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB184714 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB184714 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB184714 Supplier Novus Biologicals Supplier No. NBP282075AF647
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

LAMTOR1 Polyclonal antibody specifically detects LAMTOR1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen LAMTOR1
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias c11orf59, chromosome 11 open reading frame 59, FLJ20625, LAMTOR1, Late endosomal/lysosomal adaptor and MAPK and MTOR activator 1, late endosomal/lysosomal adaptor, MAPK and MTOR activator 1, Lipid raft adaptor protein p18, p18, p27kip1 releasing factor from RhoA, p27Kip1-releasing factor from RhoA, p27RF-RhoC11orf59, PDRO, PDROPdro, Protein associated with DRMs and endosomes, ragulator complex protein PDRO, Ragulator1, RhoA activator C11orf59
Host Species Rabbit
Immunogen Rabbit polyclonal LAMTOR1 antibody was raised against a 17 amino acid peptide near the amino terminus of human LAMTOR1 . The immunogen is located within the first 50 amino acids of LAMTOR1. Amino Acid Squence: YSSENEDSDQDREERK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 55004
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.