Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LANA2 Antibody (CM-A807), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB20167A594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB20167A594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB20167A594 Supplier Novus Biologicals Supplier No. NB200167AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LANA2 Monoclonal specifically detects LANA2 in Human, Mouse, Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockout Validated.

Specifications

Antigen LANA2
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay
Classification Monoclonal
Clone CM-A807
Conjugate Alexa Fluor 594
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockout Validated
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias LANA 2, LANA-2, Latency-associated nuclear antigen 2, vIRF 3, vIRF3, vIRF-3
Host Species Mouse
Immunogen Recombinant LANA2 fusion protein.
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity This is specific for KSHV LANA2 (residues 143-310 epitope recognition).
Target Species Human, Mouse, Virus
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.