Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LANA2 Antibody (CM-A807) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB013899 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB013899 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB013899 Supplier Novus Biologicals Supplier No. NBP280826
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LANA2 Monoclonal specifically detects LANA2 in Human, Mouse, Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunocytochemistry, Knockout Validated.

Specifications

Antigen LANA2
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay
Classification Monoclonal
Clone CM-A807
Concentration 2 mg/mL
Conjugate Unconjugated
Dilution Western Blot : 2-4 μg/mL, ELISA : 1:100-1:2000, Immunohistochemistry : 1:10-1:500, Immunocytochemistry/Immunofluorescence : 1:10-1:500, Immunoprecipitation : 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen : 1:10-1:500, Immunocytochemistry, Knockout Validated
Formulation PBS
Gene Alias LANA 2, LANA-2, Latency-associated nuclear antigen 2, vIRF 3, vIRF3, vIRF-3
Host Species Mouse
Immunogen Recombinant LANA2 fusion protein.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Reacts with viral LANA2 protein and shows no cross-reactivity to cellular proteins. Also reacts with human and mouse viral KSHV proteins.
Target Species Human, Mouse, Virus
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.