Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LANA2 Antibody (CM-A807) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB200167SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB200167SS 0.025 mL
NB200167 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB200167SS Supplier Novus Biologicals Supplier No. NB200167SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 11 publications

LANA2 Monoclonal specifically detects LANA2 in Human, Mouse, Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockout Validated.

Specifications

Antigen LANA2
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay
Classification Monoclonal
Clone CM-A807
Concentration 2.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2-4 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Knockout Validated reported in scientific literature (Golas et al)
Gene Alias LANA 2, LANA-2, Latency-associated nuclear antigen 2, vIRF 3, vIRF3, vIRF-3
Host Species Mouse
Immunogen Recombinant LANA2 fusion protein.
Purification Method Protein G purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Chromatin Research, Transcription Factors and Regulators, Virology Bacteria and Parasites
Primary or Secondary Primary
Test Specificity This is specific for KSHV LANA2 (residues 143-310 epitope recognition).
Target Species Human, Mouse, Virus
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.