Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LANCL2 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB342165 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB342165 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB342165 Supplier Novus Biologicals Supplier No. NBP281845MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LANCL2 Polyclonal antibody specifically detects LANCL2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen LANCL2
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias G protein-coupled receptor 69B, GPR69B, LanC (bacterial lantibiotic synthetase component C)-like 2, LanC lantibiotic synthetase component C-like 2 (bacterial), lanC-like protein 2, TASPMGC87139, Testis-specific adriamycin sensitivity protein
Host Species Rabbit
Immunogen LANCL2 antibody was raised against a 16 amino acid peptide from near the amino terminus of human LANCL2. The immunogen is located within amino acids 30 - 80 of LANCL2. Amino Acid Squence: VRPPATTDEPGLPFH
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 55915
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.