Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LANCL2 Antibody (OTI2A11), HRP, Novus Biologicals™
SDP

Catalog No. NBP272053H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272053H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272053H Supplier Novus Biologicals Supplier No. NBP272053H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LANCL2 Monoclonal specifically detects LANCL2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen LANCL2
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2A11
Conjugate HRP
Dilution Western Blot, Immunohistochemistry
Formulation PBS with No Preservative
Gene Alias G protein-coupled receptor 69B, GPR69B, LanC (bacterial lantibiotic synthetase component C)-like 2, LanC lantibiotic synthetase component C-like 2 (bacterial), lanC-like protein 2, TASPMGC87139, Testis-specific adriamycin sensitivity protein
Gene Symbols LANCL2
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 212-450 of human LANCL2 (NP_061167) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 55915.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.