Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LANCL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572884
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572884 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572884 Supplier Invitrogen™ Supplier No. PA572884
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LANCL2 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Necessary for abscisic acid (ABA) binding on the cell membrane and activation of the ABA signaling pathway in granulocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LANCL2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene LANCL2
Gene Accession No. Q9JJK2, Q9NS86
Gene Alias 1700003F10Rik; G protein-coupled receptor 69B; GPR69B; LanC (bacterial lantibiotic synthetase component C)-like 2; LanC lantibiotic synthetase component C-like 2; LanC lantibiotic synthetase component C-like 2 (bacterial); LanC like 2; LANCL2; lanC-like protein 2; Tasp; Testis-specific adriamycin sensitivity protein
Gene Symbols LANCL2
Host Species Rabbit
Immunogen LANCL2 antibody for a 16 amino acid peptide from near the amino terminus of human LANCL2. The immunogen is within amino acids 30 - 80 of LANCL2.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 362375, 55915, 71835
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.