Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Langerin/CD207 Antibody (306G9), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. DDX0368A488 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DDX0368A488 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DDX0368A488 Supplier Novus Biologicals Supplier No. DDX0368A488100
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Langerin/CD207 Monoclonal specifically detects Langerin/CD207 in Human, Mouse, Pig, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry.

Specifications

Antigen Langerin/CD207
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Classification Monoclonal
Clone 306G9
Concentration 0.5mg/mL
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 1:10-1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:2000
Gene Alias CD207 antigen, CD207 antigen, langerin, CD207 molecule, langerin, CLEC4KLangerhans cell specific c-type lectin, C-type lectin domain family 4 member K, C-type lectin domain family 4, member K, Langerin
Gene Symbols CD207
Host Species Mouse
Immunogen Langerin/CD207 was used as immunogen.
Purification Method Ion exchange chromatography
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 50489
Target Species Human, Mouse, Pig, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.